• USA Home
  • L3387 - L-Leucine 4-methoxy-β-naphthylamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
L3387 Sigma

L-Leucine 4-methoxy-β-naphthylamide hydrochloride

Purchase

Properties

InChI Key   NECOEODWTUWPBP-RSAXXLAASA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?