• USA Home
  • L3389 - Lectin from Psophocarpus tetragonolobus (winged bean)

EMAIL THIS PAGE TO A FRIEND
L3389 Sigma

Lectin from Psophocarpus tetragonolobus (winged bean)

TRITC conjugate, lyophilized powder

Purchase

Description

Analysis Note

Agglutination activity is expressed in μg per mL and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25 °C.

Biochem/physiol Actions

Psophocarpus tetragonolobus agglutinin is not blood group specific, but has an affinity for N-acetyl-D-galactosamine. Conjugates are prepared from purified lectin.

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing phosphate buffer salts and NaCl

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?