• USA Home
  • L3406 - pre-Liposomes Formulation 6

EMAIL THIS PAGE TO A FRIEND
L3406 Sigma

pre-Liposomes Formulation 6

Purchase

Properties

impurities   endotoxin, tested
storage temp.   −20°C

Description

Application

For preparation of neutral surface liposomes with high melting lipids:

Other Notes

L-α-phosphatidylcholine, distearoyl, 9.3 μmol
cholesterol, 6.9 μmol

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?