• USA Home
  • L3781 - pre-Liposomes Formulation 3

EMAIL THIS PAGE TO A FRIEND
L3781 Sigma

pre-Liposomes Formulation 3

Purchase

Properties

impurities   endotoxin, tested
storage temp.   −20°C

Description

Application

For preparation of negative surface charged liposomes with low melting lipids.

Other Notes

L-α-phosphatidylcholine, β-oleoyl-γ-palmitoyl, 11.9 μmol
L-α-phosphatidyl-DL-glycerol, dioleoyl, 1.3 μmol

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?