• USA Home
  • L3900 - Lauric acid N-hydroxysuccinimide ester

EMAIL THIS PAGE TO A FRIEND
L3900 Sigma

Lauric acid N-hydroxysuccinimide ester

≥98% (TLC)

Synonym: Succinimidyl laurate

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acyls, Fatty Esters, Lipids, Other Fatty Esters More...
InChI Key   LRKGVVJOUNKTGG-UHFFFAOYSA-N
assay   ≥98% (TLC)
storage temp.   −20°C

Description

Packaging

5 g in poly bottle

Biochem/physiol Actions

Lauric acid N-hydroxysuccinimide ester may be used for the synthesis of N-acylamino acids and lipidization of polypeptides or other applications that use N-hydroxysuccinimide ester conjugation chemistry.

Price and Availability


Cross-Coupling Guide

The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?