• USA Home
  • L4018 - Lectin from Lens culinaris (lentil)

EMAIL THIS PAGE TO A FRIEND
L4018 Sigma

Lectin from Lens culinaris (lentil)

agarose conjugate, saline suspension

Synonym: Lens culinaris agglutinin, LcH

Purchase

Properties

Related Categories Lens culinaris (Lentil), Affinity Chromatography, Carbohydrate Binding Matrices, Glycan Recognizing Proteins, Glycobiology,
form   saline suspension
extent of labeling   2-4 mg per mL
matrix   4% beaded agarose
matrix activation   cyanogen bromide
matrix spacer   1 atom
conjugate   agarose conjugate
storage temp.   2-8°C

Description

Biochem/physiol Actions

LcH is not blood group specific. It has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. LcH is comprised of two isomers, LcH-A (mol. wt. 49,000) and LcH-B (mol. wt. 49,000). LcH-A is mitogenic. Conjugates are prepared from purified lectin.

Physical form

Suspension in 0.9% NaCl, 1 mM CaCl2, 1 mM MnCl2 containing 0.01% thimerosal

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?