• USA Home
  • L4022 - Ricin A chain from Ricinus communis (castor bean)

EMAIL THIS PAGE TO A FRIEND
L4022 Sigma

Ricin A chain from Ricinus communis (castor bean)

deglycosylated, buffered aqueous glycerol solution

Purchase

Properties

Related Categories Ricinus communis (Castor bean), Cell Biology, Cell Signaling and Neuroscience, Glycan Recognizing Proteins, Glycobiology,
form   buffered aqueous glycerol solution
potency   ≥250 μg/mL per mL agglutination activity
composition   Protein, 0.5-3.0 mg/mL Lowry-TCA
storage temp.   2-8°C

Description

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 7.2, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hour incubation at 25°C.

Biochem/physiol Actions

Ricin A chain inactivates the 60 S ribosomal subunit of eukaryotic cells, inhibiting protein synthesis. This toxic activity is independent of the presence of the B chain of ricin.

The carbohydrate moiety of Ricin A chain has been modified using a metaperiodate-cyanoborohydride mixture. The product still inhibits protein synthesis in cell-free systems, and its removal from the blood stream by the liver is markedly decreased. Immunotoxins prepared from deglycosylated Ricin A chain will therefore be more effective in killing target cells.

Other Notes

Ricinus communis toxin (also named RCA60, RCAII, Ricin D, or RCL III) is a highly toxic protein, mol. wt. ~60 kDa, composed of two chains, A and B, connected by a single disulfide bridge.

Physical form

Solution in 40% glycerol containing 10 mM phosphate, pH 6.0, 0.15 M NaCl, 10 mM galactose and 0.5 mM dithioerythritol

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3172 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?