EMAIL THIS PAGE TO A FRIEND
L4027 Sigma

Lithium sulfate

Photographic Grade

Purchase

Properties

InChI Key   INHCSSUBVCNVSK-UHFFFAOYSA-L
quality   Photographic Grade
mp   845 °C(lit.)
density   2.22 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
OJ6419000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?