• USA Home
  • L4038 - [des-Gly10, D-Ser4, D-Trp6, Pro9]-LH-RH ethylamide trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
L4038 Sigma

[des-Gly10, D-Ser4, D-Trp6, Pro9]-LH-RH ethylamide trifluoroacetate salt

>98% (HPLC)

  • Empirical Formula (Hill Notation) C64H83N17O12

  • Molecular Weight 1282.45

  •  MDL number MFCD03452952

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, LH-RH, Neuropeptides, Peptides and Proteins,
assay   >98% (HPLC)
storage temp.   −20°C
Gene Information   human ... GNRH1(2796), GNRHR(2798)
mouse ... GNRH1(14714), GNRHR(14715)
rat ... GNRH1(25194), GNRHR(81668)

Description

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Trp-Leu-Arg-Pro-NHEt

Biochem/physiol Actions

Glp-His-Trp-Ser-Tyr-Trp-Leu-Arg-Pro-NHEt ([des-Gly10, D-Ser4, D-Trp6, Pro9]-LH-RH) is a peptide from GnRH (gonadotropin-releasing hormone) agonists. GnRH (Glp-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2), which is also referred to as LHRH (luteinizing hormone-releasing hormone) or gonadorelin, is crucial for mammalian reproduction and is released from hypothalamic neurons. It is responsible for the secretion of gonadotropins, luteinizing hormone (LH) and follicle-stimulating hormone (FSH), from the pituitary glands. Mutations in GnRH are associated with normosmic congenital hypogonadotropic hypogonadism.

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?