• USA Home
  • L4143 - Lectin from Lens culinaris (lentil)

EMAIL THIS PAGE TO A FRIEND
L4143 Sigma

Lectin from Lens culinaris (lentil)

biotin conjugate, lyophilized powder

Synonym: Lens culinaris agglutinin, LcH

Purchase

Properties

Related Categories Lens culinaris (Lentil), Glycan Recognizing Proteins, Glycobiology, Lectins, Molecular Biology,
form   lyophilized powder
composition   Protein, ~85% Lowry
extent of labeling   2-5 mol biotin per mol lectin
conjugate   biotin conjugate
storage temp.   −20°C

Description

Biochem/physiol Actions

LcH is not blood group specific. It has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. LcH is comprised of two isomers, LcH-A (mol. wt. 49,000) and LcH-B (mol. wt. 49,000). LcH-A is mitogenic. Conjugates are prepared from purified lectin.

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing sodium citrate

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?