• USA Home
  • L4511 - Lectin from Glycine max (soybean)

EMAIL THIS PAGE TO A FRIEND
L4511 Sigma

Lectin from Glycine max (soybean)

TRITC conjugate, lyophilized powder

Synonym: SBA, Soybean agglutinin

Purchase

Properties

Related Categories Glycine max (Soybean), Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent Probes, Labels, Particles and Stains, Glycan Recognizing Proteins,
form   lyophilized powder
composition   Protein, ~10% Lowry
extent of labeling   ~1 mol TRITC per mol protein
conjugate   TRITC conjugate
storage temp.   −20°C

Description

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hr incubation at 25 °C.

Biochem/physiol Actions

SBA is not blood group specific but has an affinity for N-acetyl-D-galactosamine.

Preparation Note

Conjugates are prepared from affinity purified lectin.

Physical form

Contains phosphate buffer salts and NaCl

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?