• USA Home
  • L4783 - Lipase Immobilized on cellulose from Pseudomonas sp.

EMAIL THIS PAGE TO A FRIEND
L4783 Sigma

Lipase Immobilized on cellulose from Pseudomonas sp.

≥0.5 unit/mg solid

Synonym: Triacylglycerol acylhydrolase, Triacylglycerol lipase

Purchase

Description

Biochem/physiol Actions

Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).

Other Notes

Note: When triacetin is used as substrate, the pH is 7.4. Incubation time: 60 minutes.

Unit Definition

One unit will form 1.0 μmole of ethyl octanoate from 2,2,2-trifluoroethyl octanoate per min at 25 °C.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?