• USA Home
  • L4889 - Lectin from Ulex europaeus (gorse, furze)

EMAIL THIS PAGE TO A FRIEND
L4889 Sigma

Lectin from Ulex europaeus (gorse, furze)

TRITC conjugate, lyophilized powder

Synonym: Ulex europaeus agglutinin, UEA-I

Purchase

Properties

Related Categories Ulex europaeus (Gorse, furze), Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent Probes, Labels, Particles and Stains, Glycan Recognizing Proteins,
form   lyophilized powder
potency   <10 μg/mL agglutination activity
composition   Protein, ~10% Lowry
extent of labeling   4-8 mol TRITC per mol lectin
conjugate   TRITC conjugate
storage temp.   −20°C

Description

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group O erythrocytes after 1 hour incubation at 25 °C.

Biochem/physiol Actions

Ulex europaeus agglutinin has anti-H blood group specificity. There are two types of lectin: UEA I has an affinity for L-fucose and UEA II has an affinity for N,N′-diacetylchitobiose. The mol. wt. of UEA I was initially found to be 170,000 daltons. Later reports indicate that UEA I may form aggregates at neutral and basic pH and that the correct mol. wt. is 68,000 daltons.

Linkage

Conjugates are prepared from affinity purified lectin.

Packaging

Package size based on protein content.

Physical form

Contains potassium phosphate buffer salts and sodium chloride

Price and Availability


Detect Your Unseen Protein

Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?