• USA Home
  • L4899 - Luciferase from Luciola mingrelica

EMAIL THIS PAGE TO A FRIEND
L4899 Sigma

Luciferase from Luciola mingrelica

≥85% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution

Purchase

Properties

Related Categories 1.13.x.x Acting on single donors with incorporation of molecular oxygen, 1.x.x.x Oxidoreductases, Analytical and Industrial Enzymes, Biochemicals and Reagents, Chemiluminescent compounds,
recombinant   expressed in E. coli
assay   ≥85% (SDS-PAGE)
form   buffered aqueous glycerol solution
mol wt   monomer mol wt 61,707 Da
shipped in   wet ice
storage temp.   −20°C

Description

Unit Definition

One light unit produces a biometer peak height equivalent to 0.02 μCi of 14C in PPO/POPOP cocktail. Light units measured in 50 μl assay mixture containing 5 pmol ATP and 7.5 nmol luciferin in Tris-glycine buffer, pH 7.6, at 25 °C.

Physical form

Solution in 25 mM Tris acetate, 5 mM MgSO4, 1 mM EDTA, and 50% glycerol

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?