• USA Home
  • L4908 - Lipase−polyethylene glycol

EMAIL THIS PAGE TO A FRIEND
L4908 Sigma

Lipase−polyethylene glycol

~75,000 units/mg protein (using olive oil)

Synonym: PEG-Lipase

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Enzymes, Inhibitors, and Substrates, Therapeutic Enzymes
extent of labeling   ~16 mol PEG per mol protein
storage temp.   −20°C

Description

Other Notes

Lipase from Candida rugosa coupled to methoxy-polyethylene glycol, M.W. 5,000, through secondary amine linkage.

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing PEG

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?