• USA Home
  • L5008 - L-Lactic dehydrogenase

EMAIL THIS PAGE TO A FRIEND
L5008 Sigma

L-Lactic dehydrogenase

~90% activity as H4 isoenzyme basis, ammonium sulfate suspension, ≥300 units/mg protein

Purchase

Properties

assay   ~90% activity as H4 isoenzyme basis
form   ammonium sulfate suspension
storage temp.   −20°C

Description

Analysis Note

Protein determined by biuret.

Physical form

Suspension in 3.2 M (NH4)2SO4, solution, pH 6.0

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?