• USA Home
  • L5127 - Lys-Ala dihydrobromide

EMAIL THIS PAGE TO A FRIEND
L5127 Sigma

Lys-Ala dihydrobromide

  • CAS Number 57448-86-9

  • Empirical Formula (Hill Notation) C9H19N3O3 · 2HBr

  • Molecular Weight 379.09

  •  MDL number MFCD00058268

  •  PubChem Substance ID 24896394

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   CPWAQWTXYMLNEB-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?