• USA Home
  • L5132 - L-Lactic Dehydrogenase from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
L5132 Sigma

L-Lactic Dehydrogenase from rabbit muscle

Type V-S, >95%, ammonium sulfate suspension, ≥700 units/mg protein

Synonym: (S)-Lactate: NAD+ oxidoreductase, L-LDH, LAD, LD

Purchase

Properties

Related Categories 1.1.x.x Acting on hydroxyl groups, 1.x.x.x Oxidoreductases, Application Index, Biochemicals and Reagents, Cell Biology,
type   Type V-S
assay   >95%
form   ammonium sulfate suspension
isotype   LDH5 (M4)
foreign activity   pyruvate kinase, myokinase, malic dehydrogenase, glutamic-pyruvic transaminase, glutamic-oxalacetic transaminase and α-glycerophosphate dehydrogenase <0.01%
storage temp.   2-8°C

Description

Analysis Note

Protein determined by biuret.

Biochem/physiol Actions

Also catalyzes the oxidation of other L-2-hydroxymonocarboxylic acids.

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to L-lactate per min at pH 7.5 at 37 °C.

Physical form

Crystalline suspension in 3.2 M (NH4)2SO4 solution, pH 6.0

General description

Lactic Dehydrogenase (LDH) has a total molecular weight of 140 kDa and is composed of 4 subunits which are designated M subunit (muscle) and H subunit (heart). These subunits may be mixed in any of 5 combinations (M4, M3H1, M2H2, MH3, and H4). Skeletal muscle contains LDH that is predominately M4 with some small amounts of M3H and traces of H2H2. The H and M subunits are quite similar in molecular weight, but differ substantially in amino acid composition. Rabbit muscle LDH dissociates into dimeric species (MW = ~70 kDa) in acetate-chloride at pH 5.0, the dissociation is reversible. Biochemistry, 13, 3527-3531 (1974). Oxidizes glyoxylate and lactate.
Isoelectric point: 8.4-8.6
Optimal pH : 7.5 .

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Ammonium Sulfate Suspensions - Technical Note

Note that for an ammonium sulfate suspension, most of the enzyme will be in solid form. It is likely that only negligible amounts of enzyme will be in the ammonium sulfate solution.
Keywords: Enzyme activity

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

L2500 L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?