• USA Home
  • L5259 - Luteinizing Hormone from human pituitary

EMAIL THIS PAGE TO A FRIEND
L5259 Sigma

Luteinizing Hormone from human pituitary

≥5,000 IU/mg

Synonym: ICSH, LH, PLH

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Hormones, Luteinizing Hormone (LH),
storage temp.   2-8°C
Gene Information   human ... LHB(3972), LHCGR(3973)

Description

Packaging

Package size based on RIA-active LH

Biochem/physiol Actions

Two-chain glycoprotein hormone; the α-chain is identical to that of chorionic gonadotropin, TSH, and FSH, while biological specificity is attributed to the distinct β-chain. Induces ovulation, spermatogenesis, and synthesis of steroid sex hormones. Activity is mediated through a trans-membrane LH/chorionic gonadotropin receptor that is a member of the G-protein coupled receptor (GPCR) family.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?