• USA Home
  • L5269 - Luteinizing Hormone from sheep pituitary

EMAIL THIS PAGE TO A FRIEND
L5269 Sigma

Luteinizing Hormone from sheep pituitary

lyophilized powder

Synonym: ICSH, LH, PLH

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Hormones, Luteinizing Hormone (LH),
form   lyophilized powder
storage temp.   2-8°C
Gene Information   sheep ... LHB(443395)

Description

Packaging

Package size based on NIH standard LH-525 using the ascorbic acid depletion assay. Approximately 25 units per vial.

Physical form

Lyophilized from 5 mM sodium phosphate buffer.

Biochem/physiol Actions

Two chain glycoprotein hormone, the α-chain is not active, biological specificity is attributed to the β-chain; induces ovulation, spermatogenesis, and synthesis of sex steroids.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
OK6367000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?