• USA Home
  • L5272 - Laminarinase from Trichoderma sp.

EMAIL THIS PAGE TO A FRIEND
L5272 Sigma

Laminarinase from Trichoderma sp.

powder, 100-400 units/g solid

Synonym: 1,3-[1,3;1,4]-β-D-Glucan 3(4)-glucanohydrolase, Endo-1,3(4)-β-glucanase

Purchase

Description

Other Notes

View more information on enzymes for complex carbohydrate analysis at www.sigma-aldrich.com/enzymeexplorer

Unit Definition

One unit will liberate 1.0 mg of reducing sugar (measured as glucose) from laminarin per min at pH 5.0 at 37 °C.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?