EMAIL THIS PAGE TO A FRIEND
L5522 Sigma

Levitide

≥97% (HPLC)

Synonym: pGlu-Gly-Met-Ile-Gly-Thr-Leu-Thr-Ser-Lys-Arg-Ile-Lys-Gln-NH2

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Invertebrate Neuropeptides, Neuropeptides, Peptides and Proteins,
InChI Key   LDCPZBREJGHLQM-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Other Notes

Neurohormone-like peptide found in the skin of Xenopus laevis

Price and Availability


Making Headway

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?