• USA Home
  • L5891 - Lipopolysaccharides (rough strains) from Salmonella enterica serotype typhimurium SL684 (Rc mutant)

EMAIL THIS PAGE TO A FRIEND
L5891 Sigma

Lipopolysaccharides (rough strains) from Salmonella enterica serotype typhimurium SL684 (Rc mutant)

purified by phenol-chloroform-petroleum ether extraction

Purchase

Properties

form   lyophilized powder
purified by   phenol-chloroform-petroleum ether extraction
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?