• USA Home
  • L6251 - L-Lysine Decarboxylase from Bacterium cadaveris

EMAIL THIS PAGE TO A FRIEND
L6251 Sigma

L-Lysine Decarboxylase from Bacterium cadaveris

Type III, ~0.01 unit/mg solid

Synonym: L-Lysine carboxy Lyase

Purchase

Description

Biochem/physiol Actions

Crude acetone powder from which activity can be easily extracted.

Unit Definition

One unit will release 1.0 μmole of CO2 from L-lysine per min at pH 6.0 at 37°C.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?