• USA Home
  • L6635 - Lectin from Helix aspersa (garden snail)

EMAIL THIS PAGE TO A FRIEND
L6635 Sigma

Lectin from Helix aspersa (garden snail)

lyophilized powder

Synonym: Helix aspersa agglutinin, HAA

Purchase

Properties

Related Categories Helix aspersa (Garden snail), Glycan Recognizing Proteins, Glycobiology, Lectins, Molecular Biology,
form   lyophilized powder
potency   <2 μg per mL agglutination activity
impurities   salt, free
storage temp.   −20°C

Description

Analysis Note

Where reported, agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hour incubation at 25 °C.

Biochem/physiol Actions

HAA has anti-A human blood group specificity and an affinity for terminal N-acetyl-α-D-galactosaminyl residues. Conjugates are prepared from affinity purified lectin.

Preparation Note

Affinity purified by a modification of the method of Hammarstrom and Kabat.

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?