• USA Home
  • L6761 - Lipopolysaccharides from Salmonella enterica serotype enteritidis

EMAIL THIS PAGE TO A FRIEND
L6761 Sigma

Lipopolysaccharides from Salmonella enterica serotype enteritidis

purified by trichloroacetic acid extraction

Synonym: LPS

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L, Complex Lipids,
form   lyophilized powder
purified by   trichloroacetic acid extraction
impurities   1-10% Protein (Lowry)
storage temp.   2-8°C

Description

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4).

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
OJ0895625
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?