MDL number MFCD00164307
Popular Documents: Specification Sheet (PDF)
| mol wt | mol wt 77 kDa |
| storage temp. | 2-8°C |
| Gene Information | human ... LTF(4057) |
Lactoferrin is an iron binding protein. It is structurally similar to transferrin, the plasma iron transport protein; but lactoferrin has a much higher affinity for iron (250 fold). It is very abundant in colostrum and small amounts can also be found in tears, saliva, mucous secretions and in the secondary granules of neutrophils. It is made by mucosal epithelium and neutrophils and is released by these cells in response to inflammatory stimuli. Bacterial growth is inhibited by its ability to sequester iron and also permeabilize bacterial cell walls by binding to lipopolysaccharides through its N-terminus. Lactoferrin can inhibit viral infection by binding tightly to the viral envelope protein. This prevents cell-virus fusion by blocking the binding domain. Lactoferrin appears to activate host defense systems in part by stimulating the release of interleukin-8, a neutrophil activator. It may also be involved in antibody and interleukin synthesis, lymphocyte proliferation and complement activation.
Supplied as solution in 0.05 M Sodium Acetate, pH 6.0, 0.15 M NaCl.
Customers Also Viewed
fractionated antiserum, lyophilized powder
≥85% (SDS-PAGE)
≥85% (SDS-PAGE), lyophilized powder
powder, BioReagent, suitable for cell culture
recombinant, expressed in rice, Partially iron saturated, ≥90% (SDS-PAGE)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?