EMAIL THIS PAGE TO A FRIEND
L7149 Sigma

L-Lobeline

Synonym: 2-(6-[2-Hydroxy-2-phenylethyl]-1-methyl-2-piperidinyl)-1-phenylethanone

  • CAS Number 90-69-7

  • Empirical Formula (Hill Notation) C22H27NO2

  • Molecular Weight 337.46

  •  EC Number 202-012-3

Purchase

Description

Biochem/physiol Actions

Agonist at high-affinity neuronal nicotinic acetylcholine receptors; respiratory stimulant. Inhibits synaptosomal accumulation of dopamine by interaction with the tetrabenazine binding site of the vesicular monoamine transporter (VMAT2). Blocks amphetamine-induced dopamine release.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1544 6.1/PG 3
WGK Germany 
3
RTECS 
OJ8480000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

141879 (−)-Lobeline hydrochloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?