• USA Home
  • L7400 - Lectin from Euonymus europaeus (spindle tree)

EMAIL THIS PAGE TO A FRIEND
L7400 Sigma

Lectin from Euonymus europaeus (spindle tree)

lyophilized powder

Purchase

Properties

form   lyophilized powder
potency   <1 μg per mL (using neuraminidase-treated Type B red blood cells.)
composition   Protein, ~5% Lowry
storage temp.   −20°C

Description

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions in phosphate buffered saline, pH 7.2, of a 1 mg/mL solution. This activity is the lowest concentration of lectin to agglutinate a 2% suspension of neuraminidase-treated human blood group B erythrocytes after 1 hr incubation at 25 °C.

Biochem/physiol Actions

Euonymus europaeus agglutinin (EEA) has anti-B+H blood group specificity and is reported to be most specific for blood grup B oligosaccharides having the structure α-D-Gal(1→3)[α-L-Fuc(1→2)]-β-D-Gal(1→3/4)-β-D-GlcNac, The B+H specificity is an intrinsic property of a single lectin binding site.1

Physical form

Lyophilized powder containing sodium chloride and phosphate buffer salts

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?