• USA Home
  • L7401 - Lewis-Y hexasaccharide

EMAIL THIS PAGE TO A FRIEND
L7401 Sigma

Lewis-Y hexasaccharide

from human urine

Synonym: α-Fuc-(1→2)-β-Gal-(1→4)(α-Fuc-[1→3])-β-GlcNAc-(1→3)-β-Gal-(1→4)-Glc, Lacto-N-neo-difucohexaose I, Ley-lactose

  • CAS Number 62469-99-2

  • Empirical Formula (Hill Notation) C38H65NO29

  • Molecular Weight 999.91

  •  MDL number MFCD01310882

Purchase

Properties

Related Categories Glycan Component Classes, Glycobiology, Lewis and Cell Adhesion Glycans, Molecular Biology, O-Glycans,
biological source   from human urine
InChI Key   SGJFQKVFOUQOTE-URTONSIPSA-N
storage temp.   2-8°C

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

O-Glycans

O-Linked Neutral Glycans O-Linked Sialylated Glycans Blood Group Antigens Lewis and Cell Adhesion Glycans α-Gal-(1→3)-Gal Antigens
Glycobiology Analysis Manual, 2nd Edition
Keywords: Adhesion, Catalysis, Coagulation, Degradations, Environmental, Events, Gene expression, Glycosylations, Inflammation, Ligands, O-linked glycosylation, Phosphorylations, Sequences, Transcription, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?