• USA Home
  • L7405 - Latex beads, carboxylate-modified, streptavidin-labeled, undyed

EMAIL THIS PAGE TO A FRIEND
L7405 Sigma

Latex beads, carboxylate-modified, streptavidin-labeled, undyed

buffered aqueous suspension, 1.0 μm mean particle size

Purchase

Properties

Related Categories Labware, Latex Beads, Latex and Glass Beads More...
form   buffered aqueous suspension
composition   Solids, 1.0%
mean particle size   1.0 μm

Description

Physical form

Suspension in 100 mM borate pH 8.5 + 0.1% BSA + 0.05% Tween 20 + 10mM EDTA + 0.1% sodium azide

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?