• USA Home
  • L7510 - 1-O-Palmityl-rac-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
L7510 Sigma

1-O-Palmityl-rac-glycero-3-phosphocholine

~99%

Synonym: rac-Lyso-platelet activating factor, DL-α-Lysophosphatidylcholine, γ-O-hexadecyl

  • CAS Number 17364-21-5

  • Empirical Formula (Hill Notation) C24H52NO6P

  • Molecular Weight 481.65

  •  MDL number MFCD00133273

  •  PubChem Substance ID 24896479

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Ether Lipid Metabolism, Glycerophospholipids, Glycerophosphonocholines,
InChI Key   KPUNBXYTXIWURK-UHFFFAOYSA-N
assay   ~99%
storage temp.   −20°C

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?