• USA Home
  • L7637 - Laburnum alpinum acetone powder

EMAIL THIS PAGE TO A FRIEND
L7637 Sigma

Laburnum alpinum acetone powder

Purchase

Properties

Related Categories Acetone Powders, Biochemicals and Reagents, Proteins and Derivatives More...
storage temp.   2-8°C

Description

Quality

Contains an anti-H lectin. Essentially cytisine-free. (Cytisine is the toxic alkaloid of Laburnum.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?