• USA Home
  • L7646 - Anti-Human Lambda Light Chain (Bound and Free) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
L7646 Sigma

Anti-Human Lambda Light Chain (Bound and Free) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, HU-HZ, Human Lambda Secondary Antibodies and Conjugates, Primary Antibodies,
biological source   rabbit
antibody form   IgG fraction of antiserum
clone   polyclonal
form   buffered aqueous solution
application(s)   indirect ELISA: 1:100,000
  quantitative precipitin assay: 3-4 mg/mL
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Application

Anti-Human λ Light Chain (Bound and Free) antibody produced in rabbit was used to detect free immunoglobulin light chains in sera/plasma of patients with chronic renal failure. It was used as a standard in immunoaffinity columns to isolate λ chains from effluent of patients undergoing continuous ambulatory peritoneal dialysis.

Biochem/physiol Actions

Immunoglobulins (Ig) are produced by the B cells and play crucial role in host humoral immune response. A typical immunoglobulin consists of two heavy chains and two light chains. The two light chains are either κ or the λ chain. The B cells express one type of light chain during their life time.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles

Articles

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?