• USA Home
  • L7661 - L-Lysine–Ceramic HyperD F Hydrogel Composite

EMAIL THIS PAGE TO A FRIEND
L7661 Sigma

L-Lysine–Ceramic HyperD F Hydrogel Composite

aqueous ethanol suspension, 50 μm mean particle size

Purchase

Properties

Related Categories Affinity Chromatography, Amino Acid Resins, Molecular Biology, Protein Chromatography, Proteomics More...
form   aqueous ethanol suspension
matrix   Ceramic HyperD F Hydrogel Composite
mean particle size   50 μm
capacity   >20 mg/mL binding capacity (BSA)
storage temp.   2-8°C

Description

Features and Benefits

Characteristics are tolerance of very high flow rates without bed compression, and high exchange capacity. Stable to most common solvents, and pH 2-12.

Other Notes

HyperD media are highly porous, rigid ceramic beads filled with a derivatized hydrogel.

Physical form

Suspension in 1 M NaCl with 20% ethanol

Application

Ceramic HyperD media are used in protein chromatography and affinity chromatography. Ceramic HyperD media have been used in the development of a capture step of a human recombinant F(ab’)(2) produced and expressed in baculovirus-infected cells and to identify whey proteins as a model system for chromatographic separation of proteins.

Price and Availability


Detect Your Unseen Protein

Amicon Pro Purification System
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1170 3/PG 3
WGK Germany 
3
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?