EMAIL THIS PAGE TO A FRIEND
L7664 Sigma

Leukotactin-1 human

lyophilized powder

Synonym: HCC-2, LKN1, MIP-1δ, MIP-5, NCC-3

Purchase

Properties

form   lyophilized powder
mol wt   mol wt 10 kDa
storage temp.   −20°C
Gene Information   human ... CCL15(6359)

Description

Analysis Note

The biological activity is measured by its ability to chemoattract THP-1 cells.

Biochem/physiol Actions

Potent chemoattractant for neutrophils, monocytes, and lymphocytes.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?