• USA Home
  • L8270 - Monoclonal Anti-Leukotriene (C4, D4, E4) antibody produced in rat

EMAIL THIS PAGE TO A FRIEND
L8270 Sigma

Monoclonal Anti-Leukotriene (C4, D4, E4) antibody produced in rat

clone 6E7, tissue culture supernatant

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Lipids in Cell Signaling, L-LL,
biological source   rat
antibody form   tissue culture supernatant
clone   6E7, monoclonal
contains   15 mM sodium azide
application(s)   radioimmunoassay: 1:1,000
isotype   IgG1
shipped in   dry ice
storage temp.   −20°C

Description

Analysis Note

Evaluated for antibody titer, affinity, sensitivity and specificity using a dextran coated charcoal RIA with tritiated leukotriene C4.

Immunogen

Leukotriene C4-KLH

Specificity

The antibody may be used for the quantitation of the cysteinyl-containing leukotrienes (LTC4, LTD4, LTE4) by radioimmunoassay (RIA).

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?