• USA Home
  • L8398 - L-Leucine solution 152 mM (20.0 mg/ml)

EMAIL THIS PAGE TO A FRIEND
L8398 Sigma

L-Leucine solution 152 mM (20.0 mg/ml)

Purchase

Properties

storage temp.   −20°C

Description

Application

Leucine 152mM is a concentrated stock solution for addtion to media. It is 200x with respect to RPMI-1640 and Waymouth MB 752. This product is useful in studies of the function of individual amino acids in cell culture. The biological functions of L-Leucine are frequently studied together with the other two branched-chain amino acids (BCAA), L-Ile and L-Val.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
OH2850000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

I5281 L-Isoleucine, PharmaGrade, Ajinomoto, EP, JP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
V4638 L-Valine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
I7403 L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
V0513 L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?