EMAIL THIS PAGE TO A FRIEND
L8401 Sigma

LE 300

solid

Synonym: 7-Methyl-6,7,8,9,14,15-hexahydro-5H-benz[d]indolo[2,3-g]azecine

  • Empirical Formula (Hill Notation) C20H22N2

  • Molecular Weight 290.40

  •  MDL number MFCD00951284

  •  PubChem Substance ID 24278519

Purchase

Properties

Related Categories Antagonists, Cell Biology, Cell Signaling and Neuroscience, Dopaminergics, Neuroscience,
InChI Key   YEWGIGCYIAMFMA-UHFFFAOYSA-N
form   solid
solubility   DMSO: soluble26 mg/mL
  H2O: insoluble
Gene Information   human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816)
rat ... Drd1a(24316)

Description

Biochem/physiol Actions

Potent, selective D1 dopamine receptor antagonist.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?