EMAIL THIS PAGE TO A FRIEND
L8538 Sigma

β-Lumicolchicine

  • CAS Number 6901-13-9

  • Empirical Formula (Hill Notation) C22H25NO6

  • Molecular Weight 399.44

  •  EC Number 226-365-8

  • Popular Documents:  FTNMR (PDF)  

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DJ3009400

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?