• USA Home
  • L8650 - Luteinizing Hormone, β-subunit from human pituitary

EMAIL THIS PAGE TO A FRIEND
L8650 Sigma

Luteinizing Hormone, β-subunit from human pituitary

lyophilized powder

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Hormones, Luteinizing Hormone (LH),
form   lyophilized powder
impurities   <1% α-subunit
storage temp.   −20°C
Gene Information   human ... LHB(3972)

Description

Biochem/physiol Actions

The subunit responsible for the biological specificity of LH.

Physical form

Lyophilized from 0.1 mL of 0.05 M phosphate buffer, pH 7.4.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?