EMAIL THIS PAGE TO A FRIEND
L8773 Sigma

β-Lactose

suitable for component for culture media

Synonym: β-D-Gal-(1→4)-β-D-Glc

Purchase

Properties

InChI Key   GUBGYTABKSRVRQ-DCSYEGIMSA-N
assay   ≥99% total lactose basis
  ~30% α-anomer basis
suitability   suitable for component for culture media

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?