• USA Home
  • L9018 - Lipopolysaccharides from Shigella flexneri 1A

EMAIL THIS PAGE TO A FRIEND
L9018 Sigma

Lipopolysaccharides from Shigella flexneri 1A

purified by chromatography (by gel filtration)

Purchase

Properties

form   lyophilized powder
purified by   chromatography (by gel filtration)
impurities   <1% Protein
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?