EMAIL THIS PAGE TO A FRIEND
L9033 Sigma

Edamin® K

powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Casein and Other Milk Proteins, Proteins and Derivatives More...
grade   for molecular biology
form   powder
nitrogen analysis   ~3.2% amino, ~12.3% total

Description

Application

Media for use when complete solubility is not a requirement.

Legal Information

Edamin is a registered trademark of Kerry Group

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?