• USA Home
  • L9126 - L-Lactic Dehydrogenase from chicken heart

EMAIL THIS PAGE TO A FRIEND
L9126 Sigma

L-Lactic Dehydrogenase from chicken heart

Type VIII, ammonium sulfate suspension, ≥400 units/mg protein

Synonym: (S)-Lactate: NAD+ oxidoreductase, L-LDH, LAD, LD

Purchase

Description

Analysis Note

Protein determined by biuret.

Biochem/physiol Actions

Also catalyzes the oxidation of other L-2-hydroxymonocarboxylic acids.

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to L-lactate per min at pH 7.5 at 37 °C.

Physical form

Crystalline suspension in 1.3 M (NH4)2SO4, pH 6.0

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Ammonium Sulfate Suspensions - Technical Note

Note that for an ammonium sulfate suspension, most of the enzyme will be in solid form. It is likely that only negligible amounts of enzyme will be in the ammonium sulfate solution.
Keywords: Enzyme activity

Protocols

Enzymatic Assay of L-Lactic Dehydrogenase (EC 1.1.1.27)

To standardize a procedure for the assay of L-Lactic Dehydrogenase for all sources from heart muscle.
Keywords: Extinction coefficient

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?