• USA Home
  • L9262 - Lectin from Lens culinaris (lentil)

EMAIL THIS PAGE TO A FRIEND
L9262 Sigma

Lectin from Lens culinaris (lentil)

FITC conjugate, lyophilized powder

Synonym: Lens culinaris agglutinin, LcH

Purchase

Properties

Related Categories Lens culinaris (Lentil), Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent Probes, Labels, Particles and Stains, Glycan Recognizing Proteins,
form   lyophilized powder
composition   Protein, ~40% Lowry
extent of labeling   ~2 mol FITC per mol protein
conjugate   FITC conjugate
storage temp.   −20°C

Description

Biochem/physiol Actions

LcH is not blood group specific. It has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. LcH is comprised of two isomers, LcH-A (mol. wt. 49,000) and LcH-B (mol. wt. 49,000). LcH-A is mitogenic. Conjugates are prepared from purified lectin.

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing HEPES buffer salts and sodium chloride

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?