• USA Home
  • L9378 - N-Lignoceroyl-DL-dihydrosphingosine

EMAIL THIS PAGE TO A FRIEND
L9378 Sigma

N-Lignoceroyl-DL-dihydrosphingosine

~98%

Synonym: N-Tetracosanoyl-DL-dihydrosphingosine

Purchase

Properties

Related Categories Biochemicals and Reagents, Ceramides, Lipids, N-acylsphinganines (dihydroceramides), Sphingolipids More...
InChI Key   BPLYVSYSBPLDOA-GYOJGHLZSA-N
assay   ~98%
storage temp.   −20°C

Description

Application

N-Lignoceroyl-DL-dihydrosphingosine is a racemic mixture of D and L- N-lignoceroyl-dihydrosphingosine that may be used to evaluate enantomer separation and detection technologies for dihydroceramide lipids.

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?