• USA Home
  • L9658 - Lectin from Dolichos biflorus (horse gram)

EMAIL THIS PAGE TO A FRIEND
L9658 Sigma

Lectin from Dolichos biflorus (horse gram)

TRITC conjugate, lyophilized powder

Synonym: Dolichos biflorus agglutinin, DBA

Purchase

Properties

Related Categories Dolichos biflorus (Horse gram), Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent Probes, Labels, Particles and Stains, Glycan Recognizing Proteins,
form   lyophilized powder
composition   Protein, ~15% Lowry
extent of labeling   1-3 mol TRITC per mol protein
conjugate   TRITC conjugate
storage temp.   −20°C

Description

Biochem/physiol Actions

DBA has anti-A1 human blood group specificity, useful for distinguishing between A1 and A2 blood types. DBA has strong anti-Cad activity, and has an affinity for terminal N-acetyl-α-D-galactosaminyl residues. Conjugates are prepared from affinity purified lectin.

Physical form

Contains phosphate buffer salts and NaCl

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?