• USA Home
  • L9884 - Lectin from Triticum vulgaris (wheat)

EMAIL THIS PAGE TO A FRIEND
L9884 Sigma

Lectin from Triticum vulgaris (wheat)

Evans blue conjugate, lyophilized powder

Synonym: WGA, Wheat germ agglutinin

Purchase

Properties

Related Categories Triticum vulgaris (Wheat), Glycan Recognizing Proteins, Glycobiology, Lectins, Molecular Biology,
form   lyophilized powder
composition   Protein, ~85%
extent of labeling   ~2 mol Evans blue per mol protein
conjugate   Evans blue conjugate
storage temp.   −20°C

Description

Biochem/physiol Actions

WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.

Packaging

Package size based on protein content.

Preparation Note

Conjugates are prepared from affinity purified lectin.

Physical form

Contains citrate buffer salts

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?