• USA Home
  • L9906 - Lipoprotein concentrate

EMAIL THIS PAGE TO A FRIEND
L9906 Sigma

Lipoprotein concentrate

pH 7.0-8.6

Purchase

Properties

sterility   sterile-filtered
composition   Cholesterol, 8-11 g/L
  Protein, 9-20 g/L (Biuret)
impurities   IgG, none detected
  endotoxin, level given on the label
pH   7.0-8.6
storage temp.   2-8°C

Description

Quality

Low endotoxin

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?